• USA Home
  • 594393 - 5-Ethoxy-2-fluorophenylboronic acid

EMAIL THIS PAGE TO A FRIEND
594393 Sigma-Aldrich

5-Ethoxy-2-fluorophenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Disubstituted Aryl Boronic Acids,
mp   92-97 °C (lit.)
SMILES string   CCOc1ccc(F)c(c1)B(O)O
InChI   1S/C8H10BFO3/c1-2-13-6-3-4-8(10)7(5-6)9(11)12/h3-5,11-12H,2H2,1H3
InChI key   ISJGNUFTQTZLEZ-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

Boronic Acids - ChemFiles 2007

Most boronic acids readily undergo dehydration reactions to give a cyclic (trimer) anhydride. Our selection of boronic acids may contain varying amounts of this cyclic anhydride. Fortunately, the aci...
ChemFiles 2007, 7.7, 3.
Keywords: Coupling reactions, Dehydration reaction, Suzuki coupling

Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?