• USA Home
  • 594601 - Methyl 3,5-difluorobenzoate

EMAIL THIS PAGE TO A FRIEND
594601 Aldrich

Methyl 3,5-difluorobenzoate

97%

  • Empirical Formula (Hill Notation) C8H6F2O2

  • Molecular Weight 172.13

  •  MDL number MFCD01318160

  •  PubChem Substance ID 24881523

Purchase

Properties

assay   97%
refractive index   n20/D 1.4740 (lit.)
bp   187-189 °C(lit.)
density   1.265 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
168.8 °F
Flash Point(C) 
76 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?