• USA Home
  • 594725 - 4-Propoxy-2-methylphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
594725 Sigma-Aldrich

4-Propoxy-2-methylphenylboronic acid

95%

  • Linear Formula C3H7OC6H3(CH3)B(OH)2

  • Molecular Weight 194.04

  •  MDL number MFCD06411291

  •  PubChem Substance ID 24881532

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Disubstituted Aryl Boronic Acids,
InChI Key   MMQDERNOLILBBE-UHFFFAOYSA-N
assay   95%
mp   131-136 °C (lit.)

Description

Packaging

1 g in glass bottle

Other Notes

Contains varying amounts of anhydride.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?