EMAIL THIS PAGE TO A FRIEND
595101 Sigma-Aldrich

Barium

pieces, under oil, 99%

Purchase

Properties

InChI Key   DSAJWYNOEDNPEQ-UHFFFAOYSA-N
assay   99%
form   pieces
resistivity   50.0 μΩ-cm, 20°C
bp   1640 °C(lit.)
mp   725 °C (lit.)
density   3.6 g/mL at 25 °C (lit.)

Description

Application

Critical for the synthesis of pure Ba21M2O5HX (M = Ge, SI, Ga, In, Tl) suboxide Zintl phases.3

Promotes anchorage-independent growth and invasion of human HaCaT keratinocytes via c-SRC kinase activation

Reactant for cynthesis of heterobimetallic alkaline earth complexes

Commonly used for medicinal purposes, particularly for visualization of the gastrointestinal tract

Reactant involved in:
• Interactions with fused quartz
• Aggregation of alginate-coated iron oxide nanoparticles

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Danger
Hazard statements 
RIDADR 
UN 1400 4.3 / PGII
WGK Germany 
3
RTECS 
CQ8370000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?