• USA Home
  • 595403 - (R)-(+)-2,2′-Dimethoxy-1,1′-binaphthalene

EMAIL THIS PAGE TO A FRIEND
595403 Sigma-Aldrich

(R)-(+)-2,2′-Dimethoxy-1,1′-binaphthalene

97%

Synonym: (R)-(+)-2,2′-Dimethoxy-1,1′-binaphthyl, (R)-1,1′-Bi-2-naphthyl dimethyl ether

  • CAS Number 35294-28-1

  • Empirical Formula (Hill Notation) C22H18O2

  • Molecular Weight 314.38

  •  Beilstein/REAXYS Number 4704231

  •  MDL number MFCD00091146

  •  PubChem Substance ID 24881579

Purchase

Properties

Related Categories Asymmetric Synthesis, BINOLs, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Privileged Ligands and Complexes More...
assay   97%
optical activity   [α]20/D +52°, c = 1% in chloroform
mp   227-231 °C (lit.)
InChI   1S/C22H18O2/c1-23-19-13-11-15-7-3-5-9-17(15)21(19)22-18-10-6-4-8-16(18)12-14-20(22)24-2/h3-14H,1-2H3
InChI key   BJAADAKPADTRCH-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?