• USA Home
  • 596027 - E-α-(2,4-Dichlorophenyl)-4-fluorocinnamonitrile

EMAIL THIS PAGE TO A FRIEND
596027 Sigma-Aldrich

E-α-(2,4-Dichlorophenyl)-4-fluorocinnamonitrile

97%

Purchase

Properties

Related Categories Building Blocks, C10 to C27, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds - Organic Building Blocks,
InChI Key   WDZDQWFTUNQXBS-YRNVUSSQSA-N
assay   97%
mp   142-146 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?