• USA Home
  • 596469 - (E)-4-Chloro-α-(4-fluorophenyl)cinnamonitrile

EMAIL THIS PAGE TO A FRIEND
596469 Sigma-Aldrich

(E)-4-Chloro-α-(4-fluorophenyl)cinnamonitrile

97%

Synonym: E-α-(4-Fluorophenyl)-4-chlorocinnamonitrile

Purchase

Properties

Related Categories Building Blocks, C10 to C27, Chemical Synthesis, Cyanides/Nitriles, Nitrogen Compounds - Organic Building Blocks,
assay   97%
mp   108-112 °C (lit.)
SMILES string   [H]\C(c1ccc(Cl)cc1)=C(\C#N)c2ccc(F)cc2
InChI   1S/C15H9ClFN/c16-14-5-1-11(2-6-14)9-13(10-18)12-3-7-15(17)8-4-12/h1-9H/b13-9+
InChI key   FANCAXJLECLTIF-UKTHLTGXSA-N
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?