• USA Home
  • 596612 - 2-Morpholinopyridine

EMAIL THIS PAGE TO A FRIEND
596612 Sigma-Aldrich

2-Morpholinopyridine

96%

Purchase

Properties

Related Categories Building Blocks, C9, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
assay   96%
bp   135 °C/20 mmHg (lit.)
storage temp.   room temp
SMILES string   C1CN(CCO1)c2ccccn2
InChI   1S/C9H12N2O/c1-2-4-10-9(3-1)11-5-7-12-8-6-11/h1-4H,5-8H2
InChI key   XHPVOSNOIWGRQQ-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?