• USA Home
  • 596914 - (R)-(−)-1,1′- Bi-2-naphthyl ditosylate

EMAIL THIS PAGE TO A FRIEND
596914 Sigma-Aldrich

(R)-(−)-1,1′- Bi-2-naphthyl ditosylate

98%

Purchase

Properties

Related Categories Asymmetric Synthesis, BINOLs, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, Privileged Ligands and Complexes More...
InChI Key   BKYJBVWKVKRIDN-UHFFFAOYSA-N
assay   98%
optical activity   [α]20/D -29°, c = 1% in chloroform
optical purity   ee: 97% (HPLC)
mp   176-180 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

BINOL and Derivatives

BINOL and its derivatives are one of the mostly widely used classes of ligands in asymmetric synthesis; being utilized in a broad array of reactions including: Diels–Alder, carbonyl addition and redu...
William Sommer and Daniel Weibel
Aldrich ChemFiles 2008, 8.2, 56.
Keywords: Addition reactions, Alkylations, Amidations, Asymmetric synthesis, Building blocks, Catalysis, Cycloadditions, Diels-Alder reaction, Ligands, Methods, Organic synthesis, Reductions

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?