• USA Home
  • 597805 - (3,5-Bis(trifluoromethyl)phenylethynyl)trimethylsilane

EMAIL THIS PAGE TO A FRIEND
597805 Sigma-Aldrich

(3,5-Bis(trifluoromethyl)phenylethynyl)trimethylsilane

97%

Purchase

Properties

Related Categories Alkynyl, Building Blocks, Chemical Synthesis, Halogenated Hydrocarbons, Organic Building Blocks More...
InChI Key   SKOOLTNOOKPCAJ-UHFFFAOYSA-N
assay   97%
mp   43-47 °C (lit.)

Description

Packaging

5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles

Related Content

Alkynes and Acetylenic Building Blocks

We list over 600 alkyne building blocks, including a wide array of propargyl alcohols, acetylenic boron reagents, and halogenated substrates for cross-coupling. Additionally, many of these are termin...
Keywords: Building blocks, Coupling reactions, Cross couplings

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?