• USA Home
  • 608084 - L-Histidine-13C6,α-amine-15N

EMAIL THIS PAGE TO A FRIEND
608084 Sigma-Aldrich

L-Histidine-13C6,α-amine-15N

98 atom % 15N, 99 atom % 13C

Synonym: L-Histidine-13C6,2-15N1 (α-amine-15N)

  • Empirical Formula (Hill Notation) 13C6H915NN2O2

  • Molecular Weight 162.10

  •  MDL number MFCD08702860

  •  PubChem Substance ID 329759229

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Alphabetical Listings, Amino Acids & Derivatives, G-H, Histidine, Stable Isotopes More...
isotopic purity   99 atom % 13C
  98 atom % 15N
optical activity   [α]23/D -38.4°, c = 3 in H2O
mp   282 °C (lit.)
mass shift   M+7
SMILES string   [15NH2][13C@@H]([13CH2][13c]1[13cH][nH][13cH]n1)[13C](O)=O
InChI   1S/C6H9N3O2/c7-5(6(10)11)1-4-2-8-3-9-4/h2-3,5H,1,7H2,(H,8,9)(H,10,11)/t5-/m0/s1/i1+1,2+1,3+1,4+1,5+1,6+1,7+1
InChI key   HNDVDQJCIGZPNO-HUEJSTCGSA-N

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
1

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?