• USA Home
  • 608750 - Potassium nitrate-14N

EMAIL THIS PAGE TO A FRIEND
608750 Sigma-Aldrich

Potassium nitrate-14N

99.95 atom % 14N

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Alphabetical Listings, Metabolic Labeling, Metabolomics / Proteomics, P, Stable Isotopes More...
description   15N-depleted
isotopic purity   99.95 atom % 14N
mp   334 °C (lit.)
mass shift   depleted
SMILES string   [K+].[O-][14N+]([O-])=O
InChI   1S/K.NO3/c;2-1(3)4/q+1;-1/i;1+0
InChI key   FGIUAXJPYTZDNR-QMVMUTFZSA-N

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Symbol 
GHS03  GHS03
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 1486 5.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?