• USA Home
  • 608785 - Ammonium nitrate-14N2 solution

EMAIL THIS PAGE TO A FRIEND
608785 Sigma-Aldrich

Ammonium nitrate-14N2 solution

99.95 atom % 14N

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service.
Purchase

Properties

Related Categories Agriculture, Stable Isotopes More...
description   15N-depleted
  15N-depleted
InChI Key   PRORZGWHZXZQMV-UHFFFAOYSA-N
isotopic purity   99.95 atom % 14N
bp   210 °C(lit.)
mp   169 °C (lit.)
mass shift   depleted

Description

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
RIDADR 
UN 1942 5.1 / PGIII
WGK Germany 
1
RTECS 
BR9050000

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?