• USA Home
  • 610798 - DL-Methadone-d3 solution

EMAIL THIS PAGE TO A FRIEND
610798 Supelco

DL-Methadone-d3 solution

100 μg/mL in methanol, drug standard, 98 atom % D

Synonym: DL-6-Dimethylamino-4,4-diphenyl-3-heptanone-1,1,1-d3 solution, DL-Methadone-(heptyl-1,1,1-d3) solution

  • CAS Number 60263-63-0

  • Empirical Formula (Hill Notation) C21D3H24NO

  • Molecular Weight 312.46

  •  MDL number MFCD00673285

Purchase

Properties

grade   drug standard
InChI Key   USSIQXCVUWKGNF-FIBGUPNXSA-N
isotopic purity   98 atom % D
drug control   regulated under CDSA - not available from Sigma-Aldrich Canada
availability   not available in Japan
concentration   100 μg/mL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
mass shift   M+3
storage temp.   −20°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN1230 - class 3 - PG 2 - Methanol
WGK Germany 
2
Flash Point(F) 
48.2 °F
Flash Point(C) 
9 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

HPLC Analysis of Methadone Enantiomers and Methadone Metabolite EDDP on Astec® CYCLOBOND® I 2000 HP-RSP

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Methadone Enantiomers and Methadone Metabolite EDDP on Astec® CYCLOBOND® I 2000 HP-RSP
Keywords: Analgesics, Chromatography, Clinical, Forensic, High performance liquid chromatography, Pharmaceutical

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?