• USA Home
  • 61196 - Copper(I) iodide trimethylphosphite complex

EMAIL THIS PAGE TO A FRIEND
61196 Sigma-Aldrich

Copper(I) iodide trimethylphosphite complex

≥99.0% (RT)

Synonym: Cuprous iodide trimethylphosphite complex

Purchase

Properties

Related Categories Catalysis and Inorganic Chemistry, Chemical Synthesis, Copper Catalysts More...
InChI Key   MXHFJMDNOFWFFV-UHFFFAOYSA-M
assay   ≥99.0% (RT)
mp   192-193 °C (dec.)
  192-193 °C (lit.)

Description

Other Notes

Preparation of copper enolates; Alkylation of trimethylsilylvinyl aluminates.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?