• USA Home
  • 614386 - Hydroquinone-d4-(ring-d4)

EMAIL THIS PAGE TO A FRIEND
614386 Sigma-Aldrich

Hydroquinone-d4-(ring-d4)

98 atom % D

Purchase

Properties

Related Categories Alphabetical Listings, G-H, Stable Isotopes More...
isotopic purity   98 atom % D
mass shift   M+4
SMILES string   [2H]c1c([2H])c(O)c([2H])c([2H])c1O
InChI   1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H/i1D,2D,3D,4D
InChI key   QIGBRXMKCJKVMJ-RHQRLBAQSA-N

Description

Packaging

1 g in glass bottle

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?