• USA Home
  • 630764 - 7,8-Dihydroxycoumarin

EMAIL THIS PAGE TO A FRIEND
630764 Sigma-Aldrich

7,8-Dihydroxycoumarin

90%

Synonym: Daphnetin, Daphnetol, Ruixiangsu

Purchase
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
GN6382600

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

D5564 7,8-Dihydroxycoumarin, ≥97%, powder

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?