• USA Home
  • 631604 - (R)-(+)-6,6′-Dibromo-2,2′-bis(methoxymethoxy)-1,1′-binaphthalene

EMAIL THIS PAGE TO A FRIEND
631604 Sigma-Aldrich

(R)-(+)-6,6′-Dibromo-2,2′-bis(methoxymethoxy)-1,1′-binaphthalene

97%

Purchase

Properties

InChI Key   UPLLZDVWXACTEG-UHFFFAOYSA-N
assay   97%
optical activity   [α]20/D +19.5°, c = 1% in chloroform
mp   129-133 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Indanols

These chiral imine-ligands have recently been used in the vanadium-catalyzed oxidation of sulfides in conjunction with hydrogen peroxide. Optically active sulfoxides are obtained in high yields and e...
Aldrich ChemFiles 2005, 5.4, 7.
Keywords: Asymmetric synthesis, Catalysis, Chemfiles, Ligands, Oxidations

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?