• USA Home
  • 632953 - 5,5′-Dihexyl-2,2′-bithiophene

EMAIL THIS PAGE TO A FRIEND
632953 Sigma-Aldrich

5,5′-Dihexyl-2,2′-bithiophene

96%

Synonym: DH-2T

Purchase

Properties

assay   96%
mp   29-33 °C (lit.)
SMILES string   CCCCCCc1ccc(s1)-c2ccc(CCCCCC)s2
InChI   1S/C20H30S2/c1-3-5-7-9-11-17-13-15-19(21-17)20-16-14-18(22-20)12-10-8-6-4-2/h13-16H,3-12H2,1-2H3
InChI key   RVQZMWDOMFSYKE-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles

Articles

Flexible Organic Transistors for Biomedical Applications

Tsuyoshi Sekitani,* Kazunori Kuribara, Tomoyuki Yokota, Takao Someya Department of Electric and Electronic Engineering, Department of Applied Physics, School of Engineering University of Tokyo, 7-3-1...
Keywords: Absorption, Building blocks, Degradations, Diffraction, Electronics, Evaporation, Microscopy, Nucleic acid annealing, Oxidations, Reductions, Semiconductor, Solar cells, Spectroscopy, Sterilizations, X-Ray diffraction

Related Content

Oligothiophene Synthesis

Oligothiophenes and their functional derivatives are used in a number of high-technology applications including OLEDs, OFETs and OPVs, where the ability to functionalize the oligothiophene backbone a...
Keywords: Building blocks, Coupling reactions, Sublimation, Suzuki coupling

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?