• USA Home
  • 634441 - N-Boc-3-aminomethylpyridine

EMAIL THIS PAGE TO A FRIEND
634441 Sigma-Aldrich

N-Boc-3-aminomethylpyridine

97%

Synonym: 3-[(N-(tert-Butyloxycarbonyl)amino)methyl]pyridine

Purchase

Properties

Related Categories Building Blocks, C11, Chemical Synthesis, Heterocyclic Building Blocks, Pyridines More...
InChI Key   PHQXGCNECRISKB-UHFFFAOYSA-N
assay   97%
refractive index   n20/D 1.5080(lit.)
bp   130 °C/0.5 mmHg(lit.)
density   1.041 g/mL at 25 °C(lit.)

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993BE 3 / PGII
WGK Germany 
3
Flash Point(F) 
71.6 °F
Flash Point(C) 
22 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?