EMAIL THIS PAGE TO A FRIEND
638218 Sigma-Aldrich

4-Bromopolystyrene

50-100 mesh, extent of labeling: 2.0-3.0 mmol/g loading, 1 % cross-linked with divinylbenzene

Purchase

Properties

crosslinking   1 % cross-linked with divinylbenzene
extent of labeling   2.0-3.0 mmol/g loading
particle size   50-100 mesh
storage temp.   2-8°C

Description

Packaging

50 g in glass bottle

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?