• USA Home
  • 638625 - 5-Benzoyl-2-(triisopropylsilyl)oxazole

EMAIL THIS PAGE TO A FRIEND
638625 Sigma-Aldrich

5-Benzoyl-2-(triisopropylsilyl)oxazole

97%

Purchase

Properties

assay   97%
mp   51-55 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?