• USA Home
  • 638749 - 3-Chloro-4,5-dimethoxybenzoic acid

EMAIL THIS PAGE TO A FRIEND
638749 Sigma-Aldrich

3-Chloro-4,5-dimethoxybenzoic acid

97%

Purchase

Properties

Related Categories Building Blocks, C9, Carbonyl Compounds, Carboxylic Acids, Chemical Synthesis,
InChI Key   WUBQRSNEXSSLFX-UHFFFAOYSA-N
assay   97%
mp   189-193 °C (lit.)

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?