• USA Home
  • 638927 - 2,3-Dichloro-6-nitroquinoxaline

EMAIL THIS PAGE TO A FRIEND
638927 Sigma-Aldrich

2,3-Dichloro-6-nitroquinoxaline

97%

  • CAS Number 2379-60-4

  • Empirical Formula (Hill Notation) C8H3Cl2N3O2

  • Molecular Weight 244.03

  •  MDL number MFCD00454905

  •  PubChem Substance ID 24882990

Purchase

Properties

assay   97%
mp   151-155 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?