• USA Home
  • 639567 - 3-Bromo-2-(3′-bromobenzyloxy)phenylboronic acid

EMAIL THIS PAGE TO A FRIEND
639567 Sigma-Aldrich

3-Bromo-2-(3′-bromobenzyloxy)phenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Disubstituted Aryl Boronic Acids,
InChI Key   JWOHGYXQGYSJMX-UHFFFAOYSA-N
mp   107-112 °C (lit.)

Description

Packaging

1 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles

Articles

Boronic Acids - ChemFiles 2007

Most boronic acids readily undergo dehydration reactions to give a cyclic (trimer) anhydride. Our selection of boronic acids may contain varying amounts of this cyclic anhydride. Fortunately, the aci...
ChemFiles 2007, 7.7, 3.
Keywords: Coupling reactions, Dehydration reaction, Suzuki coupling

Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?