• USA Home
  • 639702 - trans-2,3-Dimethoxy-β-nitrostyrene

EMAIL THIS PAGE TO A FRIEND
639702 Sigma-Aldrich

trans-2,3-Dimethoxy-β-nitrostyrene

97%

Synonym: 1,2-Dimethoxy-3-(2-nitrovinyl)benzene, 2-(2,3-Dimethoxyphenyl)nitroethene

Purchase

Properties

Related Categories Acyclic, Alkenes, Building Blocks, Chemical Synthesis, Organic Building Blocks More...
InChI Key   OFJZSDMKKDTNHZ-VOTSOKGWSA-N
assay   97%
mp   80-84 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis


Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?