• USA Home
  • 641847 - 2-Methylnaphtho[2,1-d]thiazole

EMAIL THIS PAGE TO A FRIEND
641847 Sigma-Aldrich

2-Methylnaphtho[2,1-d]thiazole

96%

Synonym: 2-Methyl-α-naphthothiazole

Purchase

Properties

Related Categories Building Blocks, C12 to C20+, Chemical Synthesis, Heterocyclic Building Blocks, Thiazoles More...
InChI Key   OCRIUIGNOKGMOR-UHFFFAOYSA-N
assay   96%
mp   55-61 °C
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?