• USA Home
  • 64197 - 3-(Diethoxymethylsilyl)propyl methacrylate

EMAIL THIS PAGE TO A FRIEND
64197 Sigma-Aldrich

3-(Diethoxymethylsilyl)propyl methacrylate

technical, ≥95% (GC)

Synonym: (3-Methacryloyloxypropyl)methyl-diethoxysilane, 3-(Methyldiethoxysilyl)propyl methacrylate

Purchase

Properties

grade   technical
assay   ≥95% (GC)
refractive index   n20/D 1.433
density   0.965 g/mL at 20 °C (lit.)
storage temp.   2-8°C
SMILES string   CCO[Si](C)(CCCOC(=O)C(C)=C)OCC
Safety & Documentation

Safety Information

RIDADR 
UN 2987 8 / PGII
WGK Germany 
3
Flash Point(F) 
197.6 °F
Flash Point(C) 
92 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?