• USA Home
  • 642231 - 4-[2-(4-Bromophenylthio)ethyl]pyridine

EMAIL THIS PAGE TO A FRIEND
642231 Sigma-Aldrich

4-[2-(4-Bromophenylthio)ethyl]pyridine

97%

Synonym: 4-[2-(4-Bromophenylsulfanyl)ethyl]pyridine

  • CAS Number 62237-50-7

  • Empirical Formula (Hill Notation) C13H12BrNS

  • Molecular Weight 294.21

  •  MDL number MFCD00453972

  •  PubChem Substance ID 24883181

Purchase

Properties

assay   97%
mp   40-44 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?