• USA Home
  • 64240 - N-Methallyl-N′-methylthiourea

EMAIL THIS PAGE TO A FRIEND
64240 Sigma-Aldrich

N-Methallyl-N′-methylthiourea

Synonym: N-Methyl-N′-(2-methyl-2-propen-1-yl)thiourea

  • CAS Number 21018-38-2

  • Linear Formula H2C=C(CH3)CH2NHCSNHCH3

  • Molecular Weight 144.24

  •  Beilstein/REAXYS Number 2349725

  •  EC Number 244-150-7

  •  MDL number MFCD00012209

  •  PubChem Substance ID 24883191

Purchase

Properties

mp   60-62 °C
SMILES string   CNC(=S)NCC(C)=C
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?