• USA Home
  • 642746 - 2,3-Benzofurandicarboxylic acid

EMAIL THIS PAGE TO A FRIEND
642746 Sigma-Aldrich

2,3-Benzofurandicarboxylic acid

97%

  • CAS Number 131-76-0

  • Empirical Formula (Hill Notation) C10H6O5

  • Molecular Weight 206.15

  •  MDL number MFCD05863264

  •  PubChem Substance ID 24883220

Purchase

Properties

Related Categories Benzofurans, Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks
InChI Key   FAEMVAVNTRSKEZ-UHFFFAOYSA-N
assay   97%
mp   255-259 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?