• USA Home
  • 643491 - 2-Acetylamino-5-bromo-4-methylpyridine

EMAIL THIS PAGE TO A FRIEND
643491 Sigma-Aldrich

2-Acetylamino-5-bromo-4-methylpyridine

97%

Purchase

Properties

Related Categories Building Blocks, C7 to C8, C8, Chemical Synthesis, Halogenated Heterocycles,
InChI Key   WVJCOPBDWMWUNP-UHFFFAOYSA-N
assay   97%
mp   151-155 °C (lit.)

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?