• USA Home
  • 643726 - Phosphorus pentachloride, polymer-bound

EMAIL THIS PAGE TO A FRIEND
643726 Sigma-Aldrich

Phosphorus pentachloride, polymer-bound

macroporous, 16-50 mesh, extent of labeling: 1.0-1.5 mmol/g loading

Synonym: Phosphorus(V) chloride, polymer-bound

Purchase

Properties

Related Categories C-X Bond Formation, C-X Bond Formation (Non-Halogen), Chemical Synthesis, Other Non-Halogen Reagents for C-X Bond Formation, Solid Supported Synthesis,
form   macroporous
extent of labeling   1.0-1.5 mmol/g loading
particle size   16-50 mesh
storage temp.   2-8°C

Description

Packaging

25 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?