• USA Home
  • 645125 - N-(3-Methoxybenzylidene)-4-fluoroaniline

EMAIL THIS PAGE TO A FRIEND
645125 Aldrich

N-(3-Methoxybenzylidene)-4-fluoroaniline

96%

Synonym: 4-Fluoro-N-(3-methoxybenzylidene)aniline

Purchase

Properties

assay   96%
refractive index   n20/D 1.622
density   1.181 g/mL at 25 °C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?