• USA Home
  • 645230 - 5-Butoxy-2-fluorophenylboronic acid

EMAIL THIS PAGE TO A FRIEND
645230 Sigma-Aldrich

5-Butoxy-2-fluorophenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Disubstituted Aryl Boronic Acids,
InChI Key   GKZWJVVFPFQIDP-UHFFFAOYSA-N
mp   75-80 °C (lit.)

Description

Other Notes

Contains varying amounts of anhydride

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?