• USA Home
  • 645478 - 3-Bromo-2-(3′-bromobenzyloxy)-5-methylphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
645478 Sigma-Aldrich

3-Bromo-2-(3′-bromobenzyloxy)-5-methylphenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents,
mp   109-113 °C (lit.)
SMILES string   Cc1cc(Br)c(OCc2cccc(Br)c2)c(c1)B(O)O
InChI   1S/C14H13BBr2O3/c1-9-5-12(15(18)19)14(13(17)6-9)20-8-10-3-2-4-11(16)7-10/h2-7,18-19H,8H2,1H3
InChI key   VWXCNKUTFMOYDY-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?