• USA Home
  • 645486 - 3-Bromo-2-(2′-chlorobenzyloxy)-5-methylphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
645486 Sigma-Aldrich

3-Bromo-2-(2′-chlorobenzyloxy)-5-methylphenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents,
mp   123-128 °C (lit.)
SMILES string   Cc1cc(Br)c(OCc2ccccc2Cl)c(c1)B(O)O
InChI   1S/C14H13BBrClO3/c1-9-6-11(15(18)19)14(12(16)7-9)20-8-10-4-2-3-5-13(10)17/h2-7,18-19H,8H2,1H3
InChI key   VJOGFKWLTLTDCH-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

28982 Curcumin solution, ~0.1 % (w/v) (in ethanol with 2M HCl (99:1 v/v)), for TLC derivatization

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?