• USA Home
  • 647233 - (R)-(+)-1-Boc-2-pyrrolidinecarbonitrile

EMAIL THIS PAGE TO A FRIEND
647233 Sigma-Aldrich

(R)-(+)-1-Boc-2-pyrrolidinecarbonitrile

97%

  • CAS Number 228244-20-0

  • Empirical Formula (Hill Notation) C10H16N2O2

  • Molecular Weight 196.25

  •  MDL number MFCD06411382

  •  PubChem Substance ID 24883518

  •  NACRES NA.22

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Building Blocks, Heterocyclic Building Blocks, Pyrrolidines More...
InChI Key   MDMSZBHMBCNYNO-MRVPVSSYSA-N
assay   97%
optical activity   [α]20/D +105°, c = 1% in chloroform
mp   32-36 °C (lit.)

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?