• USA Home
  • 647519 - 7-Methyl-1,4-benzothiazin-3(4H)-one

EMAIL THIS PAGE TO A FRIEND
647519 Sigma-Aldrich

7-Methyl-1,4-benzothiazin-3(4H)-one

97%

  • CAS Number 30322-02-2

  • Empirical Formula (Hill Notation) C9H9NOS

  • Molecular Weight 179.24

  •  MDL number MFCD02660690

  •  PubChem Substance ID 24883541

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Thiazines
Quality Level   100
assay   97%
mp   207-209 °C (lit.)
SMILES string   Cc1ccc2NC(=O)CSc2c1
InChI   1S/C9H9NOS/c1-6-2-3-7-8(4-6)12-5-9(11)10-7/h2-4H,5H2,1H3,(H,10,11)
InChI key   ODJQMWDFNLNCTM-UHFFFAOYSA-N

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 3
Flash Point(F) 
Not applicable
Flash Point(C) 
Not applicable

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?