• USA Home
  • 64945 - Methyl 3-sulfolene-3-carboxylate

EMAIL THIS PAGE TO A FRIEND
64945 Sigma-Aldrich

Methyl 3-sulfolene-3-carboxylate

≥98.0% (S)

Synonym: 3-Methoxycarbonyl-2,5-dihydrothiophene-1,1-dioxide, 3-Methoxycarbonyl-3-sulfolene

  • CAS Number 67488-50-0

  • Empirical Formula (Hill Notation) C6H8O4S

  • Molecular Weight 176.19

  •  Beilstein Registry Number 1368581

  •  EC Number 266-700-5

  •  MDL number MFCD00042754

  •  PubChem Substance ID 57651997

Purchase

Properties

Related Categories Building Blocks, C4 to C6, Chemical Synthesis, Heterocyclic Building Blocks, Thiophenes More...
InChI Key   YHHZABJZHHCDNW-UHFFFAOYSA-N
assay   ≥98.0% (S)
mp   60-62 °C

Description

Other Notes

A stable precursor of 2-methoxycarbonyl-1,3-butadiene, useful for cycloadditions

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?