• USA Home
  • 651192 - 5-Bromo-3-ethoxyphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
651192 Sigma-Aldrich

5-Bromo-3-ethoxyphenylboronic acid

Purchase

Description

Packaging

1, 5 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?