• USA Home
  • 652156 - 2-[[4′-(Trifluoromethoxy)phenoxy]methyl]phenylboronic acid

EMAIL THIS PAGE TO A FRIEND
652156 Sigma-Aldrich

2-[[4′-(Trifluoromethoxy)phenoxy]methyl]phenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Monosubstituted Aryl Boronic Acids,
mp   153-167 °C (lit.)
SMILES string   OB(O)c1ccccc1COc2ccc(OC(F)(F)F)cc2
InChI   1S/C14H12BF3O4/c16-14(17,18)22-12-7-5-11(6-8-12)21-9-10-3-1-2-4-13(10)15(19)20/h1-8,19-20H,9H2
InChI key   VVFYLZIEJWOSIP-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?