• USA Home
  • 652172 - 2,6-Difluoro-3-propoxyphenylboronic acid

EMAIL THIS PAGE TO A FRIEND
652172 Sigma-Aldrich

2,6-Difluoro-3-propoxyphenylboronic acid

Purchase

Properties

Related Categories Aryl Boronic Acids, Boronic Acids, Boronic Acids and Derivatives, Chemical Synthesis, Organometallic Reagents,
InChI Key   AGDROYNYQZNERV-UHFFFAOYSA-N
form   powder
mp   91-94 °C (lit.)

Description

Packaging

1, 5 g in glass bottle

Other Notes

Contains varying amounts of anhydride

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?