• USA Home
  • 652873 - 3,5-Dimethylisoxazole-4-carboxylic acid

EMAIL THIS PAGE TO A FRIEND
652873 Sigma-Aldrich

3,5-Dimethylisoxazole-4-carboxylic acid

98%

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Isoxazoles
InChI Key   IJEUISLJVBUNRE-UHFFFAOYSA-N
assay   98%
mp   141-145 °C (lit.)

Description

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

637653 3,5-Dimethylisoxazole-4-carboxylic acid, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?