• USA Home
  • 652938 - 4-Bromo-2-chloro-6-methylaniline

EMAIL THIS PAGE TO A FRIEND
652938 Sigma-Aldrich

4-Bromo-2-chloro-6-methylaniline

puriss., 98%

Synonym: 4-Bromo-6-chloro-o-toluidine

Purchase

Properties

grade   puriss.
assay   98%
mp   38-42 °C (lit.)
  41-45 °C
SMILES string   Cc1cc(Br)cc(Cl)c1N
InChI   1S/C7H7BrClN/c1-4-2-5(8)3-6(9)7(4)10/h2-3H,10H2,1H3
InChI key   DIXGIKZIIZRFKE-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

648752 4-Bromo-2-chloro-6-methylaniline, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?