• USA Home
  • 652970 - 2-Acetylamino-5-bromopyridine

EMAIL THIS PAGE TO A FRIEND
652970 Sigma-Aldrich

2-Acetylamino-5-bromopyridine

98%

  • CAS Number 7169-97-3

  • Empirical Formula (Hill Notation) C7H7BrN2O

  • Molecular Weight 215.05

  •  MDL number MFCD00468968

  •  PubChem Substance ID 24883916

Purchase

Properties

Related Categories Building Blocks, C8 to C9, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   MJFCOXATGBYERZ-UHFFFAOYSA-N
assay   98%
mp   175-179 °C (lit.)
  175-179 °C

Description

Packaging

1, 10 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

646407 2-Acetylamino-5-bromopyridine, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?