• USA Home
  • 653098 - Methyl 4-bromo-2-methoxybenzoate

EMAIL THIS PAGE TO A FRIEND
653098 Sigma-Aldrich

Methyl 4-bromo-2-methoxybenzoate

puriss., 98%

Purchase

Properties

grade   puriss.
assay   98%
form   low-melting solid
bp   100-108 °C/0.01 mmHg (lit.)
mp   33 °C (lit.)
SMILES string   COC(=O)c1ccc(Br)cc1OC
InChI   1S/C9H9BrO3/c1-12-8-5-6(10)3-4-7(8)9(11)13-2/h3-5H,1-2H3
InChI key   WPGAGRPPDYAZAD-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
>230 °F
Flash Point(C) 
>110 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

647594 Methyl 4-bromo-2-methoxybenzoate, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?