• USA Home
  • 653306 - 1-(Ethanesulfonyl)piperazine

EMAIL THIS PAGE TO A FRIEND
653306 Sigma-Aldrich

1-(Ethanesulfonyl)piperazine

97%

  • CAS Number 62937-96-6

  • Empirical Formula (Hill Notation) C6H14N2O2S

  • Molecular Weight 178.25

  •  MDL number MFCD02093528

  •  PubChem Substance ID 24883941

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, C4 to C8, Chemical Synthesis, Heterocyclic Building Blocks, Piperazines More...
InChI Key   BIYGAOBOLDXNHM-UHFFFAOYSA-N
assay   97%
mp   69-72 °C

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?