• USA Home
  • 655775 - 1-(2-Hydroxyethyl)-4-piperidone ethylene ketal

EMAIL THIS PAGE TO A FRIEND
655775 Sigma-Aldrich

1-(2-Hydroxyethyl)-4-piperidone ethylene ketal

97%

Synonym: 1,4-Dioxa-8-azaspiro[4.5]decane-8-ethanol, 2-(4,4-Ethylenedioxypiperidino)ethenol

  • CAS Number 37443-73-5

  • Empirical Formula (Hill Notation) C9H17NO3

  • Molecular Weight 187.24

  •  MDL number MFCD07784370

  •  PubChem Substance ID 24884122

  •  NACRES NA.22

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Heterocyclic Building Blocks, Piperidones
assay   97%
refractive index   n20/D 1.494
bp   75-78 °C/15 mmHg
density   1.135 g/mL at 25 °C
SMILES string   OCCN1CCC2(CC1)OCCO2
InChI   1S/C9H17NO3/c11-6-5-10-3-1-9(2-4-10)12-7-8-13-9/h11H,1-8H2
InChI key   PXTUQQILCNMBLU-UHFFFAOYSA-N

Description

Packaging

1, 5 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
WGK 2
Flash Point(F) 
230.0 °F - closed cup
Flash Point(C) 
> 110 °C - closed cup

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?