• USA Home
  • 65674 - (S)-1-{(SP)-2-[2-(Diphenylphosphino)phenyl]ferrocenyl}ethyldiphenylphosphine

EMAIL THIS PAGE TO A FRIEND
65674 Sigma-Aldrich

(S)-1-{(SP)-2-[2-(Diphenylphosphino)phenyl]ferrocenyl}ethyldiphenylphosphine

≥97%

Synonym: (1R)-1-[(1S)-1-(Diphenylphosphino)ethyl]-2-[2-(diphenylphosphino)phenyl]ferrocene (acc to CAS), Walphos SL-W002-2

Purchase

Properties

assay   ≥97%
optical purity   ee: ≥99%
SMILES string   [Fe].[CH]1[CH][CH][CH][CH]1.C[C@H]([C]2[CH][CH][CH][C]2c3ccccc3P(c4ccccc4)c5ccccc5)P(c6ccccc6)c7ccccc7
InChI   1S/C37H31P2.C5H5.Fe/c1-29(38(30-17-6-2-7-18-30)31-19-8-3-9-20-31)34-26-16-27-35(34)36-25-14-15-28-37(36)39(32-21-10-4-11-22-32)33-23-12-5-13-24-33;1-2-4-5-3-1;/h2-29H,1H3;1-5H;/t29-;;/m1../s1
InChI key   UOZGNUVEBIDUPV-SYXKTQFYSA-N

Description

General description

sold in collaboration with Solvias AG

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?